CAS RN 41011-01-2
IUPAC Name:
2-bromo-1-(3-chlorophenyl)ethan-1-one
Synonyms:
2-bromo-1-(3-chlorophenyl)ethanone
Molecular Weight (g/mol):
233.49
Molecular Formula:
C8H6BrClO
InChi Key:
KJVRURZDIOVSSQ-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC(=C1)C(=O)CBr
1
–
1
of
1
results
1
Promotions Available
2-Bromo-3'-chloroacetophenone, 97%, Thermo Scientific Chemicals
2-Bromo-3'-chloroacetophenone, 97%, Thermo Scientific Chemicals
CAS: 41011-01-2 Molecular Formula: C8H6BrClO Molecular Weight (g/mol): 233.489 MDL Number: MFCD00792669 InChI Key: KJVRURZDIOVSSQ-UHFFFAOYSA-N Synonym: 2-bromo-1-3-chlorophenyl ethanone,3-chlorophenacyl bromide,2-bromo-3'-chloroacetophenone,2-bromo-1-3-chlorophenyl ethan-1-one,2-bromo-1-3-chlorophenyl-1-ethanone,chlorophenacyl bromide,3-chlorophenacylbromide,3'-chlorophenacyl bromide,unii-n62718dtbs,ethanone, 2-bromo-1-3-chlorophenyl PubChem CID: 38738 IUPAC Name: 2-bromo-1-(3-chlorophenyl)ethanone SMILES: C1=CC(=CC(=C1)Cl)C(=O)CBr