accessibility menu, dialog, popup

UserName

FFFFFI

CAS RN 40723-80-6

IUPAC Name: 1,1,1,2,2-pentafluoro-4-iodobutane
Synonyms: 1,1,1,2,2-pentafluoro-4-iodobutane
Molecular Weight (g/mol): 273.97
Molecular Formula: C4H4F5I
InChi Key: PPXVPFPTMXFUDE-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)CCI
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results