CAS RN 407-97-6
IUPAC Name:
1-bromo-5-fluoropentane
Synonyms:
1-bromo-5-fluoropentane
Molecular Weight (g/mol):
169.04
Molecular Formula:
C5H10BrF
InChi Key:
GMYIZICPHREVDH-UHFFFAOYSA-N
SMILES:
FCCCCCBr
1
–
1
of
1
results
1
Promotions Available
1-Bromo-5-fluoropentane, 99%
1-Bromo-5-fluoropentane, 99%
CAS: 407-97-6 Molecular Formula: C5H10BrF Molecular Weight (g/mol): 169.037 MDL Number: MFCD01709395 InChI Key: GMYIZICPHREVDH-UHFFFAOYSA-N Synonym: pentane, 1-bromo-5-fluoro,5-fluoroamyl bromide,5-fluoropentyl bromide,fluoropentyl bromide,5-bromopentyl fluoride,acmc-20a2uo,1-bromo-5-fluoro-pentane,pentane,1-bromo-5-fluoro,4-01-00-00313 beilstein handbook reference,ksc493m6b PubChem CID: 120236 IUPAC Name: 1-bromo-5-fluoropentane SMILES: C(CCF)CCBr