accessibility menu, dialog, popup

UserName

CAS RN 404001-62-3

IUPAC Name: 1-hexadecyl-3-methyl-1H-imidazol-3-ium hydrate chloride
Synonyms: 3-hexadecyl-1-methylimidazol-1-ium hydrate chloride
Molecular Weight (g/mol): 361.01
Molecular Formula: C20H41ClN2O
InChi Key: YCRGEJFZTMXLMF-UHFFFAOYSA-M
SMILES: O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[N+](C)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results