accessibility menu, dialog, popup

UserName

CH3CH3H3CH3C

CAS RN 4032-86-4

IUPAC Name: 3,3-dimethylheptane
Synonyms: heptane, 3,3-dimethyl-
Molecular Weight (g/mol): 128.26
Molecular Formula: C9H20
InChi Key: BVAKDOXCVSMKHE-UHFFFAOYSA-N
SMILES: CCCCC(C)(C)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results