CAS RN 4026-18-0
IUPAC Name:
calcium bis((2S)-2-hydroxy-3-methylbutanoate)
Synonyms:
calcium bis((S)-2-hydroxy-3-methylbutyrate)
Molecular Weight (g/mol):
274.33
Molecular Formula:
C10H18CaO6
InChi Key:
CLWNJPHDLVMMEU-SCGRZTRASA-L
SMILES:
[Ca++].CC(C)[C@H](O)C([O-])=O.CC(C)[C@H](O)C([O-])=O
1
–
1
of
1
results
1
Promotions Available
2-Hydroxy-3-methylbutyric acid, 97%
2-Hydroxy-3-methylbutyric acid, 97%
CAS: 4026-18-0 Molecular Formula: C5H10O3 Molecular Weight (g/mol): 118.132 MDL Number: MFCD00004242 InChI Key: NGEWQZIDQIYUNV-UHFFFAOYSA-N Synonym: 2-hydroxy-3-methylbutyric acid,2-hydroxyisovaleric acid,alpha-hydroxyisovaleric acid,2-oxyisovaleric acid,3-methyl-2-hydroxybutyric acid,butanoic acid, 2-hydroxy-3-methyl,2-hydroxyisopentanoic acid,dl-2-hydroxyisovaleric acid,dl-2-hydroxy-3-methylbutanoic acid,2-hydroxyisopentanoate PubChem CID: 99823 ChEBI: CHEBI:60645 IUPAC Name: 2-hydroxy-3-methylbutanoic acid SMILES: CC(C)C(C(=O)O)O