accessibility menu, dialog, popup

UserName

H3COONCO

CAS RN 40203-94-9

IUPAC Name: methyl 2-isocyanato-3-phenylpropanoate
Synonyms: methyl 2-isocyanato-3-phenylpropanoate
Molecular Weight (g/mol): 205.21
Molecular Formula: C11H11NO3
InChi Key: JOTMMIYKEOOTNZ-UHFFFAOYNA-N
SMILES: COC(=O)C(CC1=CC=CC=C1)N=C=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results