accessibility menu, dialog, popup

UserName

CAS RN 4016-11-9

IUPAC Name: 2-(ethoxymethyl)oxirane
Synonyms: ethyl glycidyl ether
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: HQCSZRIVJVOYSU-UHFFFAOYNA-N
SMILES: CCOCC1CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results