accessibility menu, dialog, popup

UserName

CAS RN 40106-58-9

IUPAC Name: 3-chloro-8-thia-4,6-diazatricyclo[7.5.0.0²,⁷]tetradeca-1(9),2,4,6-tetraene
Synonyms: 3-chloro-8-thia-4,6-diazatricyclo[7.5.0.0²,⁷]tetradeca-1(9),2,4,6-tetraene
Molecular Weight (g/mol): 238.73
Molecular Formula: C11H11ClN2S
InChi Key: AUYMUMFPLSANID-UHFFFAOYSA-N
SMILES: ClC1=C2C3=C(CCCCC3)SC2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results