accessibility menu, dialog, popup

UserName

CAS RN 4004-05-1

POOOOOOO(R)HOH2NCH3CH3(Z)(Z)

CAS RN 4004-05-1

IUPAC Name: (2-aminoethoxy)[(2R)-2,3-bis[(9Z)-octadec-9-enoyloxy]propoxy]phosphinic acid
Synonyms: 1,2-dioleoyl-sn-gl dioleoyl phosphatidylethanolamine
Molecular Weight (g/mol): 744.05
Molecular Formula: C41H78NO8P
InChi Key: MWRBNPKJOOWZPW-NYVOMTAGSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OC[C@H](COP(O)(=O)OCCN)OC(=O)CCCCCCC\C=C/CCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results