CAS RN 39319-11-4
IUPAC Name:
triphenylphosphane
Synonyms:
triphenylphosphine
Molecular Weight (g/mol):
262.29
Molecular Formula:
C18H15P
InChi Key:
RIOQSEWOXXDEQQ-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
2
of
2
results
1
Promotions Available
Triphenylphosphine resin, 1% crossl. with DVB, 1.0-1.5 mmol/g, 100-200 mesh
Triphenylphosphine resin, 1% crossl. with DVB, 1.0-1.5 mmol/g, 100-200 mesh
CAS: 39319-11-4 Molecular Formula: C18H15P Molecular Weight (g/mol): 262.29 MDL Number: MFCD00003043 MFCD20489348 InChI Key: RIOQSEWOXXDEQQ-UHFFFAOYSA-N PubChem CID: 11776 IUPAC Name: triphenylphosphane SMILES: C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
2
Promotions Available
Triphenylphosphine resin, 1% crossl.with DVB, 1.0-1.5 mmol/g, 200-400 mesh, Thermo Scientific Chemicals
Triphenylphosphine resin, 1% crossl.with DVB, 1.0-1.5 mmol/g, 200-400 mesh, Thermo Scientific Chemicals
CAS: 39319-11-4 Molecular Formula: C18H15P Molecular Weight (g/mol): 262.29 MDL Number: MFCD00003043 MFCD20489348 InChI Key: RIOQSEWOXXDEQQ-UHFFFAOYSA-N PubChem CID: 11776 IUPAC Name: triphenylphosphane SMILES: C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1