accessibility menu, dialog, popup

UserName

CAS RN 39216-67-6

IUPAC Name: 4-chloro-2-(morpholin-4-yl)quinazoline
Synonyms: 4-chloro-2-(morpholin-4-yl)quinazoline
Molecular Weight (g/mol): 249.7
Molecular Formula: C12H12ClN3O
InChi Key: UWAUCVLFWGAOGM-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=CC2=NC(=N1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results