accessibility menu, dialog, popup

UserName

H3CCH3H3CCH3CH3H3C

CAS RN 3905-64-4

IUPAC Name: 2,6-di-tert-butylnaphthalene
Synonyms: 2,6-di-tert-butylnaphthalene
Molecular Weight (g/mol): 240.39
Molecular Formula: C18H24
InChi Key: TZGXZNWUOXLMFL-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC2=CC=C(C=C2C=C1)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results