accessibility menu, dialog, popup

UserName

CAS RN 38597-12-5

IUPAC Name: 4-methyl-7-{[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one
Synonyms: 4-methyl-7-{[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}chromen-2-one
Molecular Weight (g/mol): 338.31
Molecular Formula: C16H18O8
InChi Key: YUDPTGPSBJVHCN-HWSQZMMLNA-N
SMILES: CC1=CC(=O)OC2=CC(O[C@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)