accessibility menu, dialog, popup

UserName

CAS RN 3834-66-0

IUPAC Name: 1-(4-fluorophenyl)-2-phenylethane-1,2-dione
Synonyms: 1-(4-fluorophenyl)-2-phenylethane-1,2-dione
Molecular Weight (g/mol): 228.22
Molecular Formula: C14H9FO2
InChi Key: JKQPFVOJZNRINB-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results