accessibility menu, dialog, popup

UserName

CAS RN 383-64-2

IUPAC Name: 1-(ethylsulfanyl)-2,2,2-trifluoroethan-1-one
Synonyms: 1-(ethylsulfanyl)-2,2,2-trifluoroethanone
Molecular Weight (g/mol): 158.14
Molecular Formula: C4H5F3OS
InChi Key: VGGUKFAVHPGNBF-UHFFFAOYSA-N
SMILES: CCSC(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results