CAS RN 383-53-9
IUPAC Name:
2-bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one
Synonyms:
2-bromo-1-[4-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol):
267.05
Molecular Formula:
C9H6BrF3O
InChi Key:
HEMROKPXTCOASZ-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C(C=C1)C(=O)CBr
1
–
1
of
1
results
1
2-Bromo-4'-(trifluoromethyl)acetophenone 95.0+%, TCI America™
CAS: 383-53-9 Molecular Formula: C9H6BrF3O Molecular Weight (g/mol): 267.05 MDL Number: MFCD00126489 InChI Key: HEMROKPXTCOASZ-UHFFFAOYSA-N Synonym: 2-bromo-1-4-trifluoromethyl phenyl ethan-1-one,4-trifluoromethyl phenacyl bromide,2-bromo-4'-trifluoromethyl acetophenone,2-bromo-1-4-trifluoromethyl phenyl ethanone,2-bromo-4'-trifluoromethyl acetophenon,4-trifluoromethylphenacyl bromide,ethanone, 2-bromo-1-4-trifluoromethyl phenyl,4'-trifluoromethyl phenacyl bromide,2-bromo-4'-trifluoromethylacetophenone PubChem CID: 321979 IUPAC Name: 2-bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one SMILES: FC(F)(F)C1=CC=C(C=C1)C(=O)CBr