CAS RN 38186-84-4
IUPAC Name:
2-chloro-5-fluoro-3-methylpyridine
Synonyms:
2-chloro-5-fluoro-3-methylpyridine
Molecular Weight (g/mol):
145.56
Molecular Formula:
C6H5ClFN
InChi Key:
FMPYGEFGXUAAKG-UHFFFAOYSA-N
SMILES:
CC1=CC(F)=CN=C1Cl
1
–
1
of
1
results
1
2-Chloro-5-fluoro-3-methylpyridine 98.0+%, TCI America™
CAS: 38186-84-4 Molecular Formula: C6H5ClFN Molecular Weight (g/mol): 145.561 MDL Number: MFCD04972725 InChI Key: FMPYGEFGXUAAKG-UHFFFAOYSA-N Synonym: 2-chloro-5-fluoro-3-picoline,2-chloro-3-methyl-5-fluoropyridine,2-chloro-5-fluoro-3-methyl-pyridine,pyridine, 2-chloro-5-fluoro-3-methyl,pubchem6220,5-fluoro-2-chloro-3-methylpyridine,5-fluoro-2-chloro-3-methyl-pyridine,2-chloro-3-methy-5-fluorolpyridine; 2-chloro-5-fluoro-3-picoline PubChem CID: 12852014 IUPAC Name: 2-chloro-5-fluoro-3-methylpyridine SMILES: CC1=CC(=CN=C1Cl)F