CAS RN 38186-82-2
CAS RN 38186-82-2
IUPAC Name:
6-chloro-5-methylpyridin-3-amine
Synonyms:
6-chloro-5-methylpyridin-3-amine
Molecular Weight (g/mol):
142.59
Molecular Formula:
C6H7ClN2
InChi Key:
VSBISZPNLZFTPG-UHFFFAOYSA-N
SMILES:
CC1=CC(N)=CN=C1Cl
1
–
1
of
1
results
1
5-Amino-2-chloro-3-methylpyridine 97.0+%, TCI America™
CAS: 38186-82-2 Molecular Formula: C6H7ClN2 Molecular Weight (g/mol): 142.59 MDL Number: MFCD03095087 InChI Key: VSBISZPNLZFTPG-UHFFFAOYSA-N Synonym: 5-amino-2-chloro-3-picoline,2-chloro-3-methyl-5-aminopyridine,5-amino-2-chloro-3-methylpyridine,3-pyridinamine, 6-chloro-5-methyl,6-chloro-5-methyl-3-pyridylamine,6-chloro-5-methyl-pyridin-3-ylamine,6-chloro-5-methylpyridine-3-amine,pubchem1152,5-methyl-6-chloropyridine-3-amine,6-chloro-5-methyl-3-pyridinamine PubChem CID: 12295607 IUPAC Name: 6-chloro-5-methylpyridin-3-amine SMILES: CC1=CC(N)=CN=C1Cl