accessibility menu, dialog, popup

UserName

CH3H3CH3CH3CNHON

CAS RN 38078-71-6

IUPAC Name: 1-hydroxy-2,2,6,6-tetramethylpiperidine-4-carbonitrile
Synonyms: 1-hydroxy-2,2,6,6-tetramethylpiperidine-4-carbonitrile
Molecular Weight (g/mol): 182.27
Molecular Formula: C10H18N2O
InChi Key: KVTOSAMSZMXLKB-UHFFFAOYSA-N
SMILES: CC1(C)CC(CC(C)(C)N1O)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results