accessibility menu, dialog, popup

UserName

CH3SiCH3H3COH2CH2C

CAS RN 38053-91-7

IUPAC Name: (buta-1,3-dien-2-yloxy)trimethylsilane
Synonyms: (buta-1,3-dien-2-yloxy)trimethylsilane
Molecular Weight (g/mol): 142.27
Molecular Formula: C7H14OSi
InChi Key: JOAPBVRQZQYKMS-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC(=C)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results