accessibility menu, dialog, popup

UserName

CH3O

CAS RN 37940-57-1

IUPAC Name: 1-{[1,1'-biphenyl]-4-yl}propan-1-one
Synonyms: 1-{[1,1'-biphenyl]-4-yl}propan-1-one
Molecular Weight (g/mol): 210.28
Molecular Formula: C15H14O
InChi Key: JMBGDQSXJHLFTO-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results