CAS RN 3761-94-2
IUPAC Name:
1-methylcycloheptan-1-ol
Synonyms:
1-methyl-cycloheptanol
Molecular Weight (g/mol):
128.22
Molecular Formula:
C8H16O
InChi Key:
XFFKAYOHINCUNU-UHFFFAOYSA-N
SMILES:
CC1(O)CCCCCC1
1
–
1
of
1
results
1
Promotions Available
1-Methylcycloheptanol, 98%, Thermo Scientific Chemicals
1-Methylcycloheptanol, 98%, Thermo Scientific Chemicals
CAS: 3761-94-2 Molecular Formula: C8H16O Molecular Weight (g/mol): 128.215 MDL Number: MFCD00021672 InChI Key: XFFKAYOHINCUNU-UHFFFAOYSA-N Synonym: 1-methylcycloheptanol,cycloheptanol, 1-methyl,1-methylcycloheptanol-1,1-methyl-cycloheptanol,methylcycloheptanol-1,1-methyl cycloheptanol,cycloheptanol,1-methyl PubChem CID: 77376 IUPAC Name: 1-methylcycloheptan-1-ol SMILES: CC1(CCCCCC1)O