accessibility menu, dialog, popup

UserName

H3CHO

CAS RN 3760-20-1

IUPAC Name: 2-ethylcyclohexan-1-ol
Synonyms: 2-ethylcyclohexanol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: CFYUBZHJDXXXQE-UHFFFAOYNA-N
SMILES: CCC1CCCCC1O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results