accessibility menu, dialog, popup

UserName

CH3OOO

CAS RN 37443-42-8

IUPAC Name: methyl oxolane-2-carboxylate
Synonyms: methyl oxolane-2-carboxylate
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: IXHZGHPQQTXOKV-UHFFFAOYNA-N
SMILES: COC(=O)C1CCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results