CAS RN 37421-04-8
IUPAC Name:
methyl(2-phenoxyethyl)amine
Synonyms:
methyl(2-phenoxyethyl)amine
Molecular Weight (g/mol):
151.21
Molecular Formula:
C9H13NO
InChi Key:
GUMBNOITRNRLCS-UHFFFAOYSA-N
SMILES:
CNCCOC1=CC=CC=C1
1
–
1
of
1
results
1
N-Methyl-2-phenoxyethylamine, 98%
CAS: 37421-04-8 Molecular Formula: C9H13NO Molecular Weight (g/mol): 151.21 MDL Number: MFCD00085690 InChI Key: GUMBNOITRNRLCS-UHFFFAOYSA-N Synonym: methyl 2-phenoxyethyl amine,n-methyl-2-phenoxyethylamine,methyl-2-phenoxy-ethyl-amine,n-methyl-2-phenoxy-ethylamin,ethylamine, n-methyl-2-phenoxy,ethanamine, n-methyl-2-phenoxy,n-methyl-n-2-phenoxyethyl amine,alpha-phenoxy-beta-methylaminoethane,acmc-1ae2y,n-methyl-2-phenoxyethylamin PubChem CID: 37732 IUPAC Name: N-methyl-2-phenoxyethanamine SMILES: CNCCOC1=CC=CC=C1