accessibility menu, dialog, popup

UserName

CAS RN 3734-67-6

NaNaCH3ONHNHNOSOOOSOOO(Z)

CAS RN 3734-67-6

IUPAC Name: disodium (3Z)-5-acetamido-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: red 2G
Molecular Weight (g/mol): 509.41
Molecular Formula: C18H13N3Na2O8S2
InChi Key: YCUAAHNYYJTUJO-PJPLBZAPSA-L
SMILES: [Na+].[Na+].CC(=O)NC1=CC(=CC2=C1C(=O)\C(=N\NC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results