CAS RN 37310-83-1
CAS RN 37310-83-1
IUPAC Name:
(9Z)-octadec-9-en-1-ol; phosphoric acid
Synonyms:
conditioner 1; phosphoric acid
Molecular Weight (g/mol):
366.48
Molecular Formula:
C18H39O5P
InChi Key:
JGVOASSDYQIBIF-KVVVOXFISA-N
SMILES:
OP(O)(O)=O.CCCCCCCC\C=C/CCCCCCCCO
1
–
1
of
1
results
1
Oleyl Phosphate (Mono- and Di- Ester mixture), TCI America™
CAS: 37310-83-1 MDL Number: MFCD00135103 Synonym: Phosphoric Acid Oleyl Ester