accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 3728-56-1

IUPAC Name: 1-ethyl-4-methylcyclohexane
Synonyms: 1-ethyl-4-methylcyclohexane
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: CYISMTMRBPPERU-UHFFFAOYSA-N
SMILES: CCC1CCC(C)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results