accessibility menu, dialog, popup

UserName

H3CH3C

CAS RN 3728-55-0

IUPAC Name: 1-ethyl-3-methylcyclohexane
Synonyms: 1-ethyl-3-methylcyclohexane
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: UDDVMPHNQKRNNS-UHFFFAOYNA-N
SMILES: CCC1CCCC(C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results