CAS RN 37245-77-5
CAS RN 37245-77-5
IUPAC Name:
ferrioazanide pentairon azanide
Synonyms:
ferrioazanide pentairon azanide
Molecular Weight (g/mol):
365.1
Molecular Formula:
Fe6H2N2
InChi Key:
ILMFDGFXPJCFQW-UHFFFAOYSA-N
SMILES:
[NH2-].[Fe].[Fe].[Fe].[Fe].[Fe].[N-]=[Fe]
1
–
1
of
1
results
1
Iron nitride
CAS: 37245-77-5 Molecular Formula: Fe6H2N2-2 Molecular Weight (g/mol): 365.1 MDL Number: MFCD00134717 InChI Key: ILMFDGFXPJCFQW-UHFFFAOYSA-N Synonym: iron nitride,-325 mesh powder,1-aza-2,3,4-triferracyclobutan-1-yliron; diferrioamine PubChem CID: 56843031 IUPAC Name: azanide;azanidylideneiron;iron SMILES: [NH2-].[N-]=[Fe].[Fe].[Fe].[Fe].[Fe].[Fe]