CAS RN 3724-16-1
IUPAC Name:
2-(pyridin-3-yl)acetamide
Synonyms:
3-pyridine acetamide
Molecular Weight (g/mol):
136.15
Molecular Formula:
C7H8N2O
InChi Key:
YDHIMEXEGOCNHU-UHFFFAOYSA-N
SMILES:
NC(=O)CC1=CN=CC=C1
1
–
1
of
1
results
1
Promotions Available
3-Pyridineacetamide, 99%
3-Pyridineacetamide, 99%
CAS: 3724-16-1 Molecular Formula: C7H8N2O Molecular Weight (g/mol): 136.154 MDL Number: MFCD08236789 InChI Key: YDHIMEXEGOCNHU-UHFFFAOYSA-N Synonym: 2-pyridin-3-yl acetamide,3-pyridineacetamide,pyridine-3-acetamide,2-3-pyridyl acetamide,2-pyridine-3-yl-acetamide,3-pyridine acetamide,3-pyridylacetamide,acmc-1cs2v,2-3-pyridinyl acetamide,2-pyridin-3-yl acetamid PubChem CID: 564767 IUPAC Name: 2-pyridin-3-ylacetamide SMILES: C1=CC(=CN=C1)CC(=O)N