CAS RN 371-34-6
IUPAC Name:
ethyl 6-aminohexanoate
Synonyms:
ethyl 6-aminohexanoate
Molecular Weight (g/mol):
159.23
Molecular Formula:
C8H17NO2
InChi Key:
NJNQDCIAOXIFTB-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCCCCN
1
–
1
of
1
results
1
Promotions Available
Ethyl 6-aminohexanoate, 98%
Ethyl 6-aminohexanoate, 98%
CAS: 371-34-6 Molecular Formula: C8H17NO2 Molecular Weight (g/mol): 159.23 MDL Number: MFCD00233495 InChI Key: NJNQDCIAOXIFTB-UHFFFAOYSA-N Synonym: 6-aminohexanoic acid ethyl ester,ethyl 6-aminocaproate,ethyl-6-aminohexanoate,acmc-20aky1,ethyl-epsilon-aminocaproate,6-aminocaproic acid ethyl ester,6-amino-hexanoic acid ethyl ester,hexanoic acid, 6-amino-, ethyl ester PubChem CID: 406972 IUPAC Name: ethyl 6-aminohexanoate SMILES: CCOC(=O)CCCCCN