accessibility menu, dialog, popup

UserName

CAS RN 3702-98-5

IUPAC Name: ethyl 4,4,4-trichloro-3-oxobutanoate
Synonyms: ethyl 4,4,4-trichloro-3-oxobutanoate
Molecular Weight (g/mol): 233.47
Molecular Formula: C6H7Cl3O3
InChi Key: XGIRWPYENXRJMZ-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C(Cl)(Cl)Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results