accessibility menu, dialog, popup

UserName

CAS RN 3697-68-5

IUPAC Name: ethyl 1-(hydroxymethyl)cyclopropane-1-carboxylate
Synonyms: ethyl 1-(hydroxymethyl)cyclopropane-1-carboxylate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: PAILVKQSHRJIPE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1(CO)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results