accessibility menu, dialog, popup

UserName

CAS RN 36953-41-0

NHOBr

CAS RN 36953-41-0

IUPAC Name: 3-bromo-1,4-dihydropyridin-4-one
Synonyms: 3-bromo-1H-pyridin-4-one
Molecular Weight (g/mol): 174
Molecular Formula: C5H4BrNO
InChi Key: JWHZKNRXTDYNJO-UHFFFAOYSA-N
SMILES: BrC1=CNC=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results