CAS RN 36890-68-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
(C6H10O2)n
InChi Key:
—
SMILES:
*-CCCCCC(=O)O-*
1
–
1
of
1
results
1
Poly(caprolactone) diol, average M.N. 2000
CAS: 36890-68-3 Molecular Formula: (C6H10O2)n MDL Number: MFCD23713083 SMILES: *-CCCCCC(=O)O-*