accessibility menu, dialog, popup

UserName

CH3OOOH3C

CAS RN 3686-55-3

IUPAC Name: methyl 2-ethoxybenzoate
Synonyms: methyl 2-ethoxybenzoate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: QAYQKAPOTVSWLS-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results