accessibility menu, dialog, popup

UserName

CAS RN 36692-49-6

IUPAC Name: methyl 3,4-diaminobenzoate
Synonyms: methyl 3,4-diaminobenzoate
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: IOPLHGOSNCJOOO-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(N)C(N)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)