accessibility menu, dialog, popup

UserName

CAS RN 366789-02-8

IUPAC Name: 5-chloro-N-({2-oxo-3-[4-(3-oxomorpholin-4-yl)phenyl]-1,3-oxazolidin-5-yl}methyl)thiophene-2-carboxamide
Synonyms: rivaroxaban
Molecular Weight (g/mol): 435.88
Molecular Formula: C19H18ClN3O5S
InChi Key: KGFYHTZWPPHNLQ-UHFFFAOYNA-N
SMILES: ClC1=CC=C(S1)C(=O)NCC1CN(C(=O)O1)C1=CC=C(C=C1)N1CCOCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results