accessibility menu, dialog, popup

UserName

CAS RN 3663-80-7

IUPAC Name: (2S)-2,3-dihydro-1,4-benzodioxine-2-carboxylate
Synonyms: (2S)-2,3-dihydro-1,4-benzodioxine-2-carboxylate
Molecular Weight (g/mol): 179.15
Molecular Formula: C9H7O4
InChi Key: HMBHAQMOBKLWRX-QMMMGPOBSA-M
SMILES: [O-]C(=O)[C@@H]1COC2=CC=CC=C2O1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)