CAS RN 36315-01-2
CAS RN 36315-01-2
IUPAC Name:
4,6-dimethoxypyrimidin-2-amine
Synonyms:
4,6-dimethoxypyrimidin-2-amine
Molecular Weight (g/mol):
155.16
Molecular Formula:
C6H9N3O2
InChi Key:
LVFRCHIUUKWBLR-UHFFFAOYSA-N
SMILES:
COC1=CC(OC)=NC(N)=N1
1
–
1
of
1
results
1
2-Amino-4,6-dimethoxypyrimidine 98.0+%, TCI America™
CAS: 36315-01-2 Molecular Formula: C6H9N3O2 Molecular Weight (g/mol): 155.157 MDL Number: MFCD00038832 InChI Key: LVFRCHIUUKWBLR-UHFFFAOYSA-N Synonym: 2-amino-4,6-dimethoxypyrimidine,2-pyrimidinamine, 4,6-dimethoxy,4,6-dimethoxy-2-pyrimidinamine,4,6-dimethoxy-2-aminopyrimidine,4,6-dimethoxy-pyrimidin-2-ylamine,2-amino-4,6-dimethoxy pyrimidine,2-amino-4,6-dimethoxy-pyrimidine,4,6-dimethoxypyrimidine-2-ylamine,maybridge1_002259,zlchem 350 PubChem CID: 118946 IUPAC Name: 4,6-dimethoxypyrimidin-2-amine SMILES: COC1=CC(=NC(=N1)N)OC