accessibility menu, dialog, popup

UserName

CH3SBrH3C

CAS RN 362513-28-8

IUPAC Name: 3-bromo-2,4-dimethyl-5-phenylthiophene
Synonyms: 3-bromo-2,4-dimethyl-5-phenylthiophene
Molecular Weight (g/mol): 267.18
Molecular Formula: C12H11BrS
InChi Key: VJMJMLJTIYJVQY-UHFFFAOYSA-N
SMILES: CC1=C(Br)C(C)=C(S1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results