accessibility menu, dialog, popup

UserName

CAS RN 361486-60-4

H3CNNH3CCH3CH3

CAS RN 361486-60-4

IUPAC Name: N4,N4'-bis(9,9-dimethyl-9H-fluoren-2-yl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Synonyms: N4,N4'-bis(9,9-dimethylfluoren-2-yl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 720.96
Molecular Formula: C54H44N2
InChi Key: VZYZZKOUCVXTOJ-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2C2=C1C=C(C=C2)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC2=C(C=C1)C1=CC=CC=C1C2(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results