accessibility menu, dialog, popup

UserName

NNNH2NNH2N

CAS RN 36023-58-2

IUPAC Name: diaminopyrazine-2,3-dicarbonitrile
Synonyms: diaminopyrazine-2,3-dicarbonitrile
Molecular Weight (g/mol): 160.14
Molecular Formula: C6H4N6
InChi Key: FTHBTDDIVWLRLP-UHFFFAOYSA-N
SMILES: NC1=NC(C#N)=C(N=C1N)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results