CAS RN 3602-55-9
IUPAC Name:
2-tert-butylcyclohexa-2,5-diene-1,4-dione
Synonyms:
2-tert-butylcyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol):
164.2
Molecular Formula:
C10H12O2
InChi Key:
NCCTVAJNFXYWTM-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC(=O)C=CC1=O
1
–
1
of
1
results
1
2-tert-Butyl-1,4-benzoquinone 98.0+%, TCI America™
CAS: 3602-55-9 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.20 MDL Number: MFCD00666928 InChI Key: NCCTVAJNFXYWTM-UHFFFAOYSA-N PubChem CID: 19211 IUPAC Name: 2-tert-butylcyclohexa-2,5-diene-1,4-dione SMILES: CC(C)(C)C1=CC(=O)C=CC1=O