CAS RN 3535-83-9
CAS RN 3535-83-9
IUPAC Name:
tetraheptylazanium iodide
Synonyms:
tetraheptylazanium iodide
Molecular Weight (g/mol):
537.7
Molecular Formula:
C28H60IN
InChi Key:
KCSOHLKZTZMKQA-UHFFFAOYSA-M
SMILES:
[I-].CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC
1
–
1
of
1
results
1
Tetraheptylammonium Iodide 98.0+%, TCI America™
CAS: 3535-83-9 Molecular Formula: C28H60IN Molecular Weight (g/mol): 537.699 MDL Number: MFCD00041984 InChI Key: KCSOHLKZTZMKQA-UHFFFAOYSA-M Synonym: tetraheptylammonium iodide,tetra-n-heptylammonium iodide,1-heptanaminium, n,n,n-triheptyl-, iodide,tetraheptylazanium iodide,1-heptanaminium, n,n,n-triheptyl-, iodide 1:1,ammonium, tetraheptyl-, iodide,acmc-1aeuw,1-heptanaminium,n,n-triheptyl-, iodide,tetraheptylammonium iodide at PubChem CID: 77071 IUPAC Name: tetraheptylazanium;iodide SMILES: CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC.[I-]