CAS RN 3521-06-0
CAS RN 3521-06-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Basic Blue 1 90.0+%, TCI America™
CAS: 3521-06-0 Molecular Formula: C23H24Cl2N2 Molecular Weight (g/mol): 399.359 MDL Number: MFCD00050260 InChI Key: GRPFBMKYXAYEJM-UHFFFAOYSA-M Synonym: Primocyanine 6GX, Brilliant Basic Cyanine, Rhoduline Blue, Setoglaucine PubChem CID: 107404 IUPAC Name: [4-[(2-chlorophenyl)-[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=CC=C3Cl.[Cl-]