accessibility menu, dialog, popup

UserName

ClOCl

CAS RN 3511-19-1

IUPAC Name: 2,3-dichlorooxolane
Synonyms: 2,3-dichlorooxolane
Molecular Weight (g/mol): 140.99
Molecular Formula: C4H6Cl2O
InChi Key: ZQHLMWUFVRLDRK-UHFFFAOYNA-N
SMILES: ClC1CCOC1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results