accessibility menu, dialog, popup

UserName

ClH3CSCH3NH2OHO

CAS RN 3493-12-7

IUPAC Name: (3-amino-3-carboxypropyl)dimethylsulfanium chloride
Synonyms: methylmethionine chloride
Molecular Weight (g/mol): 199.69
Molecular Formula: C6H14ClNO2S
InChi Key: MYGVPKMVGSXPCQ-UHFFFAOYNA-N
SMILES: [Cl-].C[S+](C)CCC(N)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results